ContactUsLinkComponent
21 COMPOSES A 150 MG/L DANS METHANOL ( Ampoules de 1 ml ) of 4-Aminobiphenyl [CAS:92-67-1] Benzidine [CAS:92-87-5] 4-Chloro-2-methylaniline [CAS:95-69-2] 2-Aminonaphthalene [CAS:91-59-8] 4-Chloroaniline [CAS:106-47-8] 4-Methoxy-1 ,3-phenyle 1 * 5 Ampoul
Catalog # CMPLB2D3.K15.ML.1
21 COMPOSES A 150 MG/L DANS METHANOL ( Ampoules de 1 ml ) of 4-Aminobiphenyl [CAS:92-67-1] Benzidine [CAS:92-87-5] 4-Chloro-2-methylaniline [CAS:95-69-2] 2-Aminonaphthalene [CAS:91-59-8] 4-Chloroaniline [CAS:106-47-8] 4-Methoxy-1 ,3-phenyle 1 * 5 Ampoul
Catalog # CMPLB2D3.K15.ML.1
Frequently Bought Together