Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Titrators, SIMPLEX

Titrators, SIMPLEX

For milk, 0 - 25° SH
Catalog # GERB4520
Supplier:   GERBER FUNKE
Titrators, SIMPLEX
Catalog # GERB4520
Supplier:  GERBER FUNKE
Titrators, SIMPLEX
Titrators, SIMPLEX

For milk, 0 - 25° SH

Price
SFr. 199.00
Log in for availability
1 * 1 ST
Product Details & Documents

These SIMPLEX titrators consist of a polyethylene bottle on a plastic base.

  • Burette with automatic zero point adjustment
  • Precision titration by button control

Used for milk and cream.

Delivery information: Supplied with one 1 ml and one 25 ml pipette, one 200 ml Erlenmeyer flask.

Specifications
  • Description:
    For milk, 0 - 25° SH
  • Seq:
    01
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..