J.T.Baker®, SPE Glass Column Processors
Product Details & Documents
These glass column processors are specially designed glass vacuum chambers that allow multiple samples to be run simultaneously. The glass chamber allows unobstructed viewing of samples at all times.
- Run multiple samples simultaneously
- Unobstructed viewing of samples
- Specially-designed, glass vacuum chambers
The SPE-12G processor allows up to 12 samples to be run simultaneously and includes 12 Luer-type fittings, 12 individual flow control stopcocks, 12 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates a variety of glassware.
The SPE-24G processor allows up to 24 samples to be processed in 20 to 30 minutes and includes 24 Luer-type fittings, 24 individual flow control stopcocks, 24 polypropylene collection needles, a vacuum gauge/controller, and a rack set that accommodates a variety of glassware.
Delivery information: Both units are supplied with a vacuum manifold, a sample collection rack with three support posts, four adjustable height shelves with support clips, and an instruction manual.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:SPE-12G column processor
- Seq:01
Accessories
Product Information
- Catalog #AvailabilityPrice
- Luer connector, male, PP
Product Information
- Catalog #AvailabilityPrice
- Luer connector, female, PP
Product Information
- Catalog #AvailabilityPrice
- Needles, stainless steel
Product Information
- Catalog #AvailabilityPrice
- Glass vacuum chamber, for SPE-24G column processorGlass vacuum chamber, for SPE-24G column processor7423-00Log in for availability1 * 1 ST - SFr. 491.00
Product Information
- Catalog #AvailabilityPrice
- Gasket seals, neoprene, for SPE-24G column processorGasket seals, neoprene, for SPE-24G column processor7435-00Log in for availability1 * 1 ST - SFr. 84.30
Product Information
- Catalog #AvailabilityPrice
- Needles, PP
Product Information
- Catalog #AvailabilityPrice
- Vacuum gauge/controller assembly
Product Information
- Catalog #AvailabilityPrice
- Luer stopcocks, stainless steel
Product Information
- Catalog #AvailabilityPrice
- Stopcocks, PP
Product Information
- Catalog #AvailabilityPrice
- Luer stopcocks, PTFE