Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 500 ml
Catalog # CAYM600228-500
Supplier:   Cayman Chemical
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Catalog # CAYM600228-500
Supplier:  Cayman Chemical
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 500 ml

CAS Number:  
Price
166.00 SFr.
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 ml
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications