Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 250 mL
Catalog # CAYM600228-250
Supplier:   Cayman Chemical
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)
Catalog # CAYM600228-250
Supplier:  Cayman Chemical
CAS Number:  
Buffer, sodium acetate buffer solution pH 5,2 (3 mol/L)

Sodium acetate buffer solution pH 5,2 (3 mol/l), Glass bottle, 250 mL

CAS Number:  
Price
SFr. 87.60
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    Sodium acetate buffer solution pH 5,2 (3 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.