Behenic acid
Glass bottle for solids, 1 g
Catalog # CAYM9000338-1
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Docosanoic acid
- CAS Number:112-85-6
- Formula:C₂₂H₄₄O₂
- MDL Number:MFCD00002807
- Molecular Weight:340.59 g/mol
- Merck Index:13,01024
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:80 °C
- Density:0.8221 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-010-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



