Lignoceric acid
Glass bottle for solids, 500 mg
CAYM13353-500
: Tetracosanoic acid
- :557-59-5
- :C₂₄H₄₈O₂
- :MFCD00002810
- :368.64 g/mol
- :13,05509
- :406 °C (1013 hPa)
- :87 °C
- :0.8207 g/cm³ (100 °C)
- :Ambient
- :209-180-7
- Pack type:Glass bottle for solids
- Pk:500 mg