Lignoceric acid
Glass bottle for solids, 250 mg
Katalognr. CAYM13353-250
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
Synonyme: Tetracosanoic acid
- CAS Nummer:557-59-5
- Formel:C₂₄H₄₈O₂
- MDL-Nummer:MFCD00002810
- Molekulargewicht:368.64 g/mol
- Merck-Index:13,05509
- Siedepunkt:406 °C (1013 hPa)
- Schmelzender Platingehalt:87 °C
- Dichte:0.8207 g/cm³ (100 °C)
- Lagertemperatur:Ambient
- EINECS:209-180-7
Technische Daten
Technische Daten
- Pack type:Glass bottle for solids
- Pk:250 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



