Lignoceric acid
Glass bottle for solids, 100 mg
Catalog # CAYM13353-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Tetracosanoic acid
- CAS Number:557-59-5
- Formula:C₂₄H₄₈O₂
- MDL Number:MFCD00002810
- Molecular Weight:368.64 g/mol
- Merck Index:13,05509
- Boiling Pt:406 °C (1013 hPa)
- Melting Pt:87 °C
- Density:0.8207 g/cm³ (100 °C)
- Storage Temperature:Ambient
- EINECS:209-180-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



