ContactUsLinkComponent
23 COMPOSES A 2MG/L DANS DICHLOROMETHANE (Ampoules de 1 ml ) of Benzo(b)fluoranthene [CAS:205-99-2] Benzo(k)fluoranthene [CAS:207-08-9] Benzo(a)pyrene [CAS:50-32-8] Indeno(1,2,3-c ,d)pyrene [CAS:193-39-5] Dibenzo(a,h)anthracene [CAS:53-70-3] 1 * 5 Ampoul
Catalog # CMPLD664.2K.DCM.1
23 COMPOSES A 2MG/L DANS DICHLOROMETHANE (Ampoules de 1 ml ) of Benzo(b)fluoranthene [CAS:205-99-2] Benzo(k)fluoranthene [CAS:207-08-9] Benzo(a)pyrene [CAS:50-32-8] Indeno(1,2,3-c ,d)pyrene [CAS:193-39-5] Dibenzo(a,h)anthracene [CAS:53-70-3] 1 * 5 Ampoul
Catalog # CMPLD664.2K.DCM.1
Frequently Bought Together