ContactUsLinkComponent
26 COMPOSES DANS CARBONE DISULFURE (Ampoules de 2 ml ) n-Pentane [CAS:109-66-0] 2500ug/ml n-Hexane [CAS:110-54-3] 2500ug/ml n-Heptane [CAS:142-82-5] 2500ug/ml n-Octane [CAS:111-65-9] 2500ug/ml n-Nonane [CAS:111-84-2] 2500ug/ml n-Decane [CAS 1 * 10 Ampoul
Catalog # CMPLFF9D.2K5.CDS.2
26 COMPOSES DANS CARBONE DISULFURE (Ampoules de 2 ml ) n-Pentane [CAS:109-66-0] 2500ug/ml n-Hexane [CAS:110-54-3] 2500ug/ml n-Heptane [CAS:142-82-5] 2500ug/ml n-Octane [CAS:111-65-9] 2500ug/ml n-Nonane [CAS:111-84-2] 2500ug/ml n-Decane [CAS 1 * 10 Ampoul
Catalog # CMPLFF9D.2K5.CDS.2
Frequently Bought Together