Shakers
Shaker and stand for thirty six Mojonnier tubes
Catalog # 614-0185
Supplier: GERBER FUNKE
Jump to:
Product Details & Documents
Shakers for determining the fat content in milk according to Mojonnier's method. White PVC housing. Choice of models to accept 4, 6 or 36 Mojonnier extraction tubes. Units provide forceful, uniform and reproducible mixing.
- Timer, 0 to 15 min
- Produce 100 shakes per minute
- Model for 36 butyrometers is supplied with stand
Specifications
- Description:Shaker and stand for thirty six Mojonnier tubes
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cork stopper for extraction tubes, Mojonnier`s methodCork stopper for extraction tubes, Mojonnier`s method614-0188Log in for availability1 * 1 ST - SFr. 0.40
Product Information
- Catalog #AvailabilityPrice
- Extraction tubes, Mojonnier, with flattened bulbExtraction tubes, Mojonnier, with flattened bulb614-0187Log in for availability1 * 1 ST - SFr. 23.40
Product Information
- Catalog #AvailabilityPrice
- Extraction tubes, Mojonnier, with rounded bulbExtraction tubes, Mojonnier, with rounded bulb614-0186Log in for availability1 * 1 ST - SFr. 23.40
Product Information
- Catalog #AvailabilityPrice
- Stand, wooden, for 12 extraction tubes acc. to Mojonnier`s methodStand, wooden, for 12 extraction tubes acc. to Mojonnier`s method614-0189Log in for availability1 * 1 ST - SFr. 85.60