ContactUsLinkComponent
MiniVaq CASE VACCUM INULATING BOX, 15 litres The MiniVaq Case is a vacuum insulated box for up to 72 h transport time. Rigid outer case, made of aluminium frames with coloured cabinet covering. Top handle for easy carrying and lifting of lid. Lid clo 1 * 1 items
Catalog # DLTTB100039DE
Supplier:  DELTA T
MiniVaq CASE VACCUM INULATING BOX, 15 litres The MiniVaq Case is a vacuum insulated box for up to 72 h transport time. Rigid outer case, made of aluminium frames with coloured cabinet covering. Top handle for easy carrying and lifting of lid. Lid clo 1 * 1 items
Catalog # DLTTB100039DE
Supplier:  DELTA T
Frequently Bought Together