Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cooling elements and frames

Cooling elements and frames

Cooling elements, −18 °C, Tempshell, Translucent white, −22...−18 °C, 275×155×60 mm
210×110×30 mm
Catalog # DLTTA100017DE
Supplier:   DELTA T
Cooling elements and frames
Catalog # DLTTA100017DE
Supplier:  DELTA T
Cooling elements and frames

Cooling elements, −18 °C, Tempshell, Translucent white, −22...−18 °C, 275×155×60 mm
210×110×30 mm

Price
SFr. 108.00
Log in for availability
1 * 1 PAIRE
Product Details & Documents

Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.

  • Made of PE and natural in colour
  • Sealing made of laminated aluminium film with a protected cap
  • A liquid crystal thermometer in the elements shows its current temperature

Used for transportation and storage of samples like blood products, homograft’s, cell cultures, stem cells, antibodies, proteins and other temperature sensitive medications.

Inside dimension: 210×110×30 mm

Specifications
  • Temperature Range:
    −22...−18 °C
  • Colour:
    Translucent white
  • Description:
    Cooling elements, −18 °C, Tempshell
  • Dimensions:
    275×155×60 mm
    210×110×30 mm
  • Seq:
    03