ContactUsLinkComponent
Support frame combi model SKM750, 18/10 stainless steel. This demonstration model is a special combination of HEAVY FRAME (26,9mm) & DIN-SUPPORT FRAME MATERIAL (12mm). It has a significant high STABILITY. The clamps, bossheads & container included o 1 * 1 items
Catalog # BOCHSKM750
Supplier:  BOCHEM
Support frame combi model SKM750, 18/10 stainless steel. This demonstration model is a special combination of HEAVY FRAME (26,9mm) & DIN-SUPPORT FRAME MATERIAL (12mm). It has a significant high STABILITY. The clamps, bossheads & container included o 1 * 1 items
Catalog # BOCHSKM750
Supplier:  BOCHEM
Frequently Bought Together