Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Base frames

Base frames

Base frame, V shape (2 legs), 600 mm, 400 mm
Catalog # BOCH5645
Supplier:   BOCHEM
Base frames
Catalog # BOCH5645
Supplier:  BOCHEM
Base frames
Base frames

Base frame, V shape (2 legs), 600 mm, 400 mm

Price
SFr. 258.00
Log in for availability
1 * 1 ST
Product Details & Documents

These tube base frames made from stainless steel 18/10 are available in "V" and "H" shape.

Specifications
  • Description:
    Base frame, V shape (2 legs)
  • Length:
    600 mm
  • Seq:
    01
  • Arm length:
    400 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.