Orders LinkContactUsLinkComponent
Agar, Bacteriological Grade
Agar, Bacteriological Grade
Bottle, 1 kg
  RC-004
Agar, Bacteriological Grade
  RC-004
 :  9002-18-0
Agar, Bacteriological Grade
Agar, Bacteriological Grade
Bottle, 1 kg
 :  9002-18-0
SFr. 430.00
1 * 1 kg
A purified agar that is a fine light colored powder that is used as a solidification agent in the preparation of microbiological culture media.
Bacteriological grade purity: Low contaminant levels suitable for sensitive microbiological applications.
 :  Agar Agar
  •  :9002-18-0
  •  :(C₁₂H₁₈O₉)n
  •  :MFCD00081288
  •  :90 °C
  • Pack type:
    Bottle
  • Pk:
    1 kg