ContactUsLinkComponent
VarioPrep HPLC column VP 250/32 NUCLEOSIL 100-5 C18 *** with adjustable Endfitting *** length: 250 mm, ID: 32 mm pack of 1 1 * 1 items
Catalog # MANA715340.320
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC column VP 250/32 NUCLEOSIL 100-5 C18 *** with adjustable Endfitting *** length: 250 mm, ID: 32 mm pack of 1 1 * 1 items
Catalog # MANA715340.320
Supplier:  MACHEREY-NAGEL
Frequently Bought Together