Watchmakers' magnifiers
Magnifier with plano-convex lens, 25 mm
Catalog # ESCH11247
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
- Includes vent to prevent condensation
- Made of flame resistant plastic material
- Colour: anthracite
Specifications
- Ø:25 mm
- Magnification:7×
- Description:Magnifier with plano-convex lens
- Seq:002