INSERT 0.35ML SHELL STYLE 1 * 1.000 items
Catalog # NSCAC4015-542
Supplier: THERMO NATIONAL SCIENTIFIC
INSERT 0.35ML SHELL STYLE 1 * 1.000 items
Catalog # NSCAC4015-542
Supplier: THERMO NATIONAL SCIENTIFIC
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Screw caps for Jerricans
Supplier: Kautex
Nitric acid 67%, NORMATOM® for trace metal analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test tubes with round or flat bottom, Corning® Gosselin™
Supplier: Corning