ContactUsLinkComponent
16 components Al 5mg/l B 5mg/l Ba 1mg/l Ca 500mg/l Co 1mg/l Cu 5mg/l Fe 5mg/l K 500mg/l Mg 500mg/l Mn 5mg/l Mo 1mg/l Na 500mg/l P 200mg/l Sr 1mg/l Zn 5mg/l Si from (NH4)2SiF6 100mg/l in HNO3 5% Conservation 12 months. o 1 * 250 mL
Catalog # CMPL95D5.5.5N
16 components Al 5mg/l B 5mg/l Ba 1mg/l Ca 500mg/l Co 1mg/l Cu 5mg/l Fe 5mg/l K 500mg/l Mg 500mg/l Mn 5mg/l Mo 1mg/l Na 500mg/l P 200mg/l Sr 1mg/l Zn 5mg/l Si from (NH4)2SiF6 100mg/l in HNO3 5% Conservation 12 months. o 1 * 250 mL
Catalog # CMPL95D5.5.5N
Frequently Bought Together