Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Spoon spatulas, micro

Spoon spatulas, micro

300 mm, 5 mm
Catalog # 231-0457
Supplier:   RSG
Spoon spatulas, micro
Catalog # 231-0457
Supplier:  RSG
Spoon spatulas, micro
Spoon spatulas, micro

300 mm, 5 mm

Price
54.20 SFr.
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

  • With a micro spoon at one end and a micro spatula at the other
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Specifications
  • Length:
    300 mm
  • Blade width:
    5 mm