VWR® UltraPure, Buffer, PBS solution (phosphate buffered saline)
Phosphate buffered saline (PBS), 1X solution, Dulbecco's formulation, Bag-in-box (Cubitainer), 20 L
Supplier: VWR Chemicals
VWR® UltraPure, Buffer, PBS solution (phosphate buffered saline)
Catalog # K812-20L
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
Dulbecco's formulation without calcium and magnesium.
- Free from DNase, RNase or protease
137,7 mM sodium chloride, 2,7 mM potassium chloride, 9,55 mM phosphate buffer
pH @ 25 °C 7,1 to 7,7
Documents
Specifications
Specifications
- Pack type:Bag-in-box (Cubitainer)
- Pk:20 L
- Description:Phosphate buffered saline (PBS), 1X solution, Dulbecco's formulation
- Cat. No.:K812-20L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



