Azotometer/saccharometer
Azotometer/saccharometer, 0 − 100 ml, 1/10
Catalog # NEUB0345-60-050105
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Azotometer/saccharometer made of borosilicate glass, with glass base.
Specifications
- Description:Azotometer/saccharometer
- Seq:01
- Division:1/10
- Range:0 − 100 ml