Orders LinkContactUsLinkComponent
Counting chambers
Counting chambers
Slides with four chambers and counting grids, with grid, 3×3 mm
Catalog # 630-1537
Supplier:   Kova International
Counting chambers
Catalog # 630-1537
Supplier:  Kova International
Counting chambers
Slides with four chambers and counting grids, with grid, 3×3 mm
Price
SFr. 221.00
Log in for availability
1 * 100 ST
Product Details & Documents
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
  • High optical quality through the composition acrylate
  • Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
  • Capillary action for reproducible precision
  • Homogeneous suspension of sample sediments: Retained volume of 1 µl
  • For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl
Specifications
  • Type:
    with grid
  • W×D:
    3×3 mm
  • Description:
    Slides with four chambers and counting grids