Multi-parameter meter (pH/mV/DO), bench, Orion Star™ A216
This benchtop meter is ideal for a wide range of applications and advanced pH and dissolved oxygen analysis in the lab. It offers two measuring channels allowing users to measure pH and dissolved oxygen simultaneously or view each channel separately. Can be used with RDO optical or polarographic DO probes.
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Measurement stability indicator and selectable read modes - Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Non volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- Three year meter warranty
IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.
Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two.
Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values and calibration editing option that allows individual points to be fixed without a full recalibration. Calibrate dissolved oxygen using water-saturated air, air-saturated water, custom value (Winkler titration) and zero point options.
Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings.
Lieferinformationen: Supplied with electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8157BNUMD ROSS Ultra Triode refillable epoxy-body pH/ATC electrode, 083005MD Polarographic DO sensor with calibration sleeve, ROSS pH buffer and storage solution kit, DO sensor maintenance kit and BOD adapter, funnel and stirrer.
- Description:Bench pH/DO meter Orion Star A216, kit incl. ROSS Ultra Triode pH/ATC probe and polarographic DO probe
- Seq:02
- Connections:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
- DO range:0,00 - 90,00 mg/L
- Battery type:4 AA (sold separately)
- Battery life:800 h
- Compliance:CE, TUV 3-1, FCC Class A
- DO concentration resolution:0,1/0,01 mg/L
- Memory:Non-volatile memory preserves data log, calibration log and meter settings
- IP rating:IP 54
- DO accuracy:Polarographic: ±0,2 mg/L or ±2% of reading, whichever is greater
RDO: ±0,1 mg/L (up to 8 mg/L), ±0,2 mg/L (8 - 20 mg/L), ±10% of reading (up to 50 mg/L) - Data logging capacity:2000 with date and time stamp
- Languages:English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
- Power supply:AC adapter (included) or 4 AA batteries
- mV resolution:0,1
- DO calibration:Water-saturated air, air saturated water, manual (Winkler) and zero point
- Input:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature), stirrer input (stirrer probe)
- W×D×H:240×180×110 mm
- Temperature range:–5,0...+50,0 °C
- Operating temperature range:5...45 °C (ambient)
- mV range:±2000,0
- DO barometric pressure range:Automatic or manual, 450 - 850 mm Hg
- No. of channels:2
- pH range:–2,000...+20,000
- Calibration:pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
- Pressure range:400 - 850 mm Hg
- Temperature compensation:Automatic or manual
- pH calibration:Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- Temperature resolution:0,1 °C
- DO concentration:0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
- Temperature accuracy:±0,1 °C
- Weight:0,9 kg
- Cat. No.:665-0274
- pH accuracy:±0,002
- pH resolution:0,1/0,01/0,001
- Data logging - log function:Manual/Automatic with AUTO-READ™
- Alarm:High/low limit alarm, calibration due alarm
- Model:A216
- Outputs:RS232, USB
- mV accuracy:±0,2 mV or ±0,05% of reading, whichever is greater
- Calibration points:1 point relative mV (RmV) mode, 1 point ORP mode
- DO saturation:0 - 600% (polarographic); 0 - 500% (RDO)
- Display:Graphic LCD with backlight
Accessories
- DescriptionFor
- ORIOSTARA-BEABenchtop meter-attached electrode stand (electrode holder arm and head), Orion™ Star A series, 360° motion (without weighted base)Orion Star A110 + A210 series, Orion Versa Star benchtop meters1 * 1 ST - SFr. 202.00
- Description
- 662-1140Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable1 * 1 SET - SFr. 94.40
- Description
- 662-1547Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters1 * 1 ST - SFr. 89.50
- Description
- ORIOSTARA-HBOrion™ Star™ A Series weighted base for use with meter-attached electrode stand1 * 1 ST - SFr. 69.60
- Description
- Description
- ORIO1010005Cable adapter, for connecting the RS232 port of a meter to a USB port1 * 1 ST - SFr. 162.00
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



