ContactUsLinkComponent
Standard rack for upright freezer for (hxd) 3x3 = 9 boxes 100mm high, delivered including standard white cardboard boxes and 9x9 cel l dividers, stainless steel, measures (hxdxw) 321x423x139mm 1 * 1 items
Catalog # TENATE23202B
Supplier:  TENAK
Standard rack for upright freezer for (hxd) 3x3 = 9 boxes 100mm high, delivered including standard white cardboard boxes and 9x9 cel l dividers, stainless steel, measures (hxdxw) 321x423x139mm 1 * 1 items
Catalog # TENATE23202B
Supplier:  TENAK
Frequently Bought Together