Probes for optima series baths and circulators
External probe, flexible plastic, 3 m cable
Catalog # 461-0630
Supplier: GRANT INSTRUMENTS
Jump to:
Product Details & Documents
Documents
GRANTXSEP_TDS_EN
Datasheet
Specifications
- Description:External probe, flexible plastic, 3 m cable
- Seq:01
- Application:Monitoring temperature of external loads