Rotary evaporating flasks
25 ml, 14/23 NS
Catalog # LENZ03041422
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Documents
Specifications
- Socket size:14/23 NS
- Capacity:25 ml
- Seq:03