ContactUsLinkComponent
SOLUTION DE 9 COMPOSES DANS HNO3 2% Ca 1000mg/l K 500mg/l Na 250mg/l P 800mg/l Mg 100mg/l Cu 2mg/l Mn 1mg/l Fe 80mg/l Zn 3mg/l 1 * 100 mL
Catalog # CMPL80F2.1K.2N.L1
SOLUTION DE 9 COMPOSES DANS HNO3 2% Ca 1000mg/l K 500mg/l Na 250mg/l P 800mg/l Mg 100mg/l Cu 2mg/l Mn 1mg/l Fe 80mg/l Zn 3mg/l 1 * 100 mL
Catalog # CMPL80F2.1K.2N.L1
Frequently Bought Together