Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stopcocks, straight bore

Stopcocks, straight bore

6 mm, 12 mm
Catalog # BIBBSST106
Supplier:   SciLabware
Stopcocks, straight bore
Catalog # BIBBSST106
Supplier:  SciLabware
Stopcocks, straight bore
Stopcocks, straight bore

6 mm, 12 mm

Price
SFr. 310.00
Log in for availability
1 * 5 ST
Product Details & Documents

PYREX®, borosilicate glass with PTFE key.

  • Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
  • Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Specifications
  • Hole Ø:
    6 mm
  • For tube I-Ø:
    12 mm
Accessories

Product Information

  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare PTFE keys for stopcocks, PYREX®, straight bore
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    BIBBSST102K
    1,5 mm
    Log in for availability
    1 * 5 ST - SFr. 124.00

Product Information

  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare PTFE keys for stopcocks, PYREX®, straight bore
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    BIBBSST103K
    2,5 mm
    Log in for availability
    1 * 5 ST - SFr. 145.00

Product Information

  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare PTFE keys for stopcocks, PYREX®, straight bore
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    BIBBSST104K
    4,0 mm
    Log in for availability
    1 * 5 ST - SFr. 206.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.