ContactUsLinkComponent
9 COMPOSES B 50mg/l Al 50mg/l Fe 50mg/l Zn 50mg/l Sr 50mg/l Ba 50mg/l Ni 10mg/l Cu 10mg/l Co 10mg/l DANS HNO3 4% 1 * 250 mL
Catalog # CMPL4A1D.50.4N
9 COMPOSES B 50mg/l Al 50mg/l Fe 50mg/l Zn 50mg/l Sr 50mg/l Ba 50mg/l Ni 10mg/l Cu 10mg/l Co 10mg/l DANS HNO3 4% 1 * 250 mL
Catalog # CMPL4A1D.50.4N
Frequently Bought Together