ContactUsLinkComponent
7 COMPOSES A 1G/L DANS H2O 1000mg/l each of Chlorides (Cl-) Nitrites (NO2-) Sulphates (SO42-) Nitrates (NO3-) Phosphates (PO43-) Fluorides (F-) Acetates (CH3COO-) 1 * 100 mL
Catalog # CMPL4F41.1K.W.L1
7 COMPOSES A 1G/L DANS H2O 1000mg/l each of Chlorides (Cl-) Nitrites (NO2-) Sulphates (SO42-) Nitrates (NO3-) Phosphates (PO43-) Fluorides (F-) Acetates (CH3COO-) 1 * 100 mL
Catalog # CMPL4F41.1K.W.L1
Frequently Bought Together