Adapters, glass, cone/screwthread
24/29 NS, GL 14, 5,5 - 6,5 mm
Katalognr. LENZ01301424
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Dokumente
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Technische Daten
- Cone size:24/29 NS
- Thread:GL 14
- Tube ext.Ø:5,5 - 6,5 mm
- Seq:03