Adapters, glass, cone/screwthread
24/29 NS, GL 25, 9 - 11 mm
Catalog # LENZ01302524
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Cone size:24/29 NS
- Thread:GL 25
- Tube ext.Ø:9 - 11 mm
- Seq:09