Orders LinkContactUsLinkComponent
Pasteur pipettes
Pasteur pipettes
—, 150 mm, —, +, Individually wrapped
Catalog # KART329
Supplier:   KARTELL
Pasteur pipettes
Catalog # KART329
Supplier:  KARTELL
Pasteur pipettes
Pasteur pipettes
—, 150 mm, —, +, Individually wrapped
Price
SFr. 157.00
Log in for availability
1 * 500 ST
Product Details & Documents
PE, disposable, sterile or non sterile.
  • Disposable PE pipettes with bulb and graduations
  • The slender tip guarantees precision transfer of the sample
  • Non sterile pipettes are available in multi-packs; sterile pipettes are individually wrapped
Caution: *Capillary
Specifications
  • Packed:
    Individually wrapped
  • Sterile:
    +
  • Capacity:
  • Length:
    150 mm
  • Seq:
    1
  • Graduation: