ContactUsLinkComponent
up to 171l/min, up to 99.5%*, without compressor, without compressor, 7-10 bar g (101.5-145 psi g) 1 * 1 items
Catalog # PARKN2-80
Supplier:  PARKER HANNIFIN
up to 171l/min, up to 99.5%*, without compressor, without compressor, 7-10 bar g (101.5-145 psi g) 1 * 1 items
Catalog # PARKN2-80
Supplier:  PARKER HANNIFIN
Frequently Bought Together