Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Transfer pipettes

Transfer pipettes

Graduated, 1,0 ml, 33 µl, 150 mm, +, Bulk
Catalog # RATI2655171
Supplier:   RATIOLAB
Transfer pipettes
Catalog # RATI2655171
Supplier:  RATIOLAB
Transfer pipettes
Transfer pipettes

Graduated, 1,0 ml, 33 µl, 150 mm, +, Bulk

Price
SFr. 92.20
Log in for availability
1 * 500 ST
Product Details & Documents

PE, disposable.

  • Soft, elastic bulb
  • Unbreakable, reduced risk of injury compared to glass
  • Inert to biological fluids and most acids
  • Graduated pipettes with clearly visible embossed graduations
  • Sterile pipettes are gas-sterilised and individually packed
  • Suitable for food contact
Specifications
  • Packed:
    Bulk
  • Sterile:
    +
  • Capacity:
    1,0 ml
  • Description:
    Graduated
  • Length:
    150 mm
  • Seq:
    04
  • Drop vol.:
    33 µl