Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Staining jars

Staining jars

Staining jar, with lid, 105×85×70 mm
Catalog # MARI4200004
Supplier:   MARIENFELD
Staining jars
Catalog # MARI4200004
Supplier:  MARIENFELD
Staining jars
Staining jars

Staining jar, with lid, 105×85×70 mm

Price
SFr. 352.00
Log in for availability
1 * 10 ST
Product Details & Documents

Staining set consists of glass dish and staining jar made of soda lime glass, and staining tray made of either pressed glass or stainless steel.

These staining tray hold up to 10 slides (76×26 mm).

Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.

Specifications
  • Description:
    Staining jar, with lid
  • W×D×H:
    105×85×70 mm