Recombinant Murine IL-7 1 * 10 µG
Catalog # PEPR217-17-10UG
Supplier: PeproTech, Inc.
Frequently Bought Together
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
MSD GOLD 96-WELL STREPTAVIDIN SECTOR PLATE PACK 1 * 5 items
Supplier: MESO SCALE DISCOVERY
VWR® Cell Culture Dishes
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Electroporation Cuvettes Plus
Supplier: BTX
Stoppers for tubes
Supplier: Greiner Bio-One
Fernbach culture flasks, Nalgene®
Supplier: Thermo Fisher Scientific
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
Plates and modules, FluoroNunc™ and LumiNunc™
Supplier: Thermo Fisher Scientific
Racks, magnetic separation, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Lids for Greiner 1536-well microplates
Supplier: Greiner Bio-One
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
PCR microtubes without caps, 0.2 ml, Amplitube™
Supplier: Simport Scientific
Mouthpieces for blood pipettes, Assistent
Supplier: GLASWARENFABRIK KARL HECHT
Optical bottom plates, SCREENSTAR
Supplier: Greiner Bio-One
S-Trap™ 96-well Mini Plate 1 * 3 items
Supplier: MARKET SOURCE PART PROCESS
Strips of 8 PCR tubes with detached cap strips
Supplier: Brand