ContactUsLinkComponent
MELANGE DE 21 COMPOSES DANS METHANOL Vinylchloride [CAS:75-01-4] 125mg/l Dichloromethane [CAS:75-09-2] 1250mg/l Chloroform [CAS:67-66-3] 1000mg/l 1,1,1-Trichloroethane [CAS:71-55-6] 125mg/l Tetrachloromethane [CAS:56-23-5] 125mg/l 1 ,2-Dich 1 * 10 Ampoul
Catalog # CMPL442F.K15.ML.1
MELANGE DE 21 COMPOSES DANS METHANOL Vinylchloride [CAS:75-01-4] 125mg/l Dichloromethane [CAS:75-09-2] 1250mg/l Chloroform [CAS:67-66-3] 1000mg/l 1,1,1-Trichloroethane [CAS:71-55-6] 125mg/l Tetrachloromethane [CAS:56-23-5] 125mg/l 1 ,2-Dich 1 * 10 Ampoul
Catalog # CMPL442F.K15.ML.1
Frequently Bought Together