ContactUsLinkComponent
ROTILABO WIDE-NECK CANS (250ML) Made of HDPE. Leakproof. With press-in plugs and screw cap. Ideal for storing powders or granulated substances. Large wide-neck opening enables easy filling and cleaning. (neck inside 50MM)(69x94MM) 1 * 50 items
Catalog # ROTHHT57.1
Supplier:  Roth Carl
ROTILABO WIDE-NECK CANS (250ML) Made of HDPE. Leakproof. With press-in plugs and screw cap. Ideal for storing powders or granulated substances. Large wide-neck opening enables easy filling and cleaning. (neck inside 50MM)(69x94MM) 1 * 50 items
Catalog # ROTHHT57.1
Supplier:  Roth Carl
Frequently Bought Together