Order Entry
Switzerland
Orders LinkContactUsLinkComponent
LED bulbs

LED bulbs

LED module 4500 K ±300 K
Catalog # 630-1320
Supplier:   MOTIC
LED bulbs
Catalog # 630-1320
Supplier:  MOTIC
LED bulbs
LED bulbs

LED module 4500 K ±300 K

Price
SFr. 120.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    LED module 4500 K ±300 K
  • Seq:
    302