Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Redox buffer solutions

Redox buffer solutions

Redox buffer, 468 mV, 0,1, 6×30 ml, 6
Catalog # METT51350064
Supplier:   Mettler - Toledo
Redox buffer solutions
Catalog # METT51350064
Supplier:  Mettler - Toledo
CAS Number:  
Redox buffer solutions

Redox buffer, 468 mV, 0,1, 6×30 ml, 6

CAS Number:  
Price
SFr. 46.80
Log in for availability
1 * 6 ST
Product Details & Documents

Redox buffer solutions are used for verification of all common redox sensors.

  • Temperature dependent
  • Used in quality assurance systems

A table on every bottle label indicates redox values at different temperatures and hence not used for calibration purposes.

 : Supplied with a quality inspection certificate.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pk:
    6
  • Description:
    Redox buffer, 468 mV
  • Pk Info:
    6×30 ml
  • pH value:
    0,1

Specifications