Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

Technical buffer solution, 250 ml, 7,00, 250 mL
Catalog # METT51350006
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METT51350006
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 250 ml, 7,00, 250 mL

CAS Number:  
Price
SFr. 31.00
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Technical buffer solution, 250 ml
  • pH value:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.