Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST/DIN

pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 6,865, 250 mL
Catalog # METT51350054
Supplier:   Mettler - Toledo
pH buffer solutions, NIST/DIN
Catalog # METT51350054
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 6,865, 250 mL

CAS Number:  
Price
SFr. 37.50
Log in for availability
1 * 250 mL
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

 : NIST traceable.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    NIST/DIN buffer solution
  • pH value:
    6,865

Specifications