Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST/DIN

pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 4,006, 250 mL
Catalog # METT51350052
Supplier:   Mettler - Toledo
pH buffer solutions, NIST/DIN
Catalog # METT51350052
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 4,006, 250 mL

CAS Number:  
Price
SFr. 37.50
Log in for availability
1 * 250 mL
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

Certifications: NIST traceable.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    NIST/DIN buffer solution
  • pH value:
    4,006

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.