Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH buffer solutions, certified (DKD)

pH buffer solutions, certified (DKD)

DKD certified buffer solution, 10,00, 1× 250 ml, 250 mL
Catalog # METT51350038
Supplier:   Mettler - Toledo
pH buffer solutions, certified (DKD)
Catalog # METT51350038
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, certified (DKD)

DKD certified buffer solution, 10,00, 1× 250 ml, 250 mL

CAS Number:  
Price
33.80 SFr.
Log in for availability
1 * 250 mL
Product Details & Documents

Certifications: The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the ideally suitable products for regulated industries. DKD is accepted by other accreditation bodies within the scope of the European Cooperation for Accreditation (EA) and the International Laboratory Accreditation Cooperation (ILAC).

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    DKD certified buffer solution
  • Pk Info:
    1× 250 ml
  • pH value:
    10,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.