Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

pH buffer solutions

Technical buffer solution, 250 ml, 2,00, 250 mL
Catalog # METT51350002
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METT51350002
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 250 ml, 2,00, 250 mL

CAS Number:  
Price
SFr. 31.00
Log in for availability
1 * 250 mL
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Technical buffer solution, 250 ml
  • pH value:
    2,00

Specifications