Examination light
Examination light, HALUX LED 20 P LX, CH-plug
Catalog # 139-0183
Supplier: WALDMANN
Jump to:
Product Details & Documents
Powerful down to the smallest details, it does not diffuse any infrared radiation - no heat. A twin-wall body including fan optimizes cooling of luminaire head. A simple control unit is integrated on luminaire head. Friction system for a risk-free and precise light positioning.
- Standard adapter for different fixture types
- Electronic transformer
- Robust and ergonomic design
- Long LED service life: Reduced maintenance costs
- Easy cable replacement: Device connector on transformer housing
IEC 60601-2-41 standard examination light
Specifications
- Description:Examination light, HALUX LED 20 P LX, CH-plug